Gene Transcripts

Gene Symbol
RAP2B
Entrez Gene ID
5912
Gene Name
RAP2B, member of RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 3: 153162212 - 153170624 on Build GRCh38
UniGene ID
Hs.700713
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002886.3 NP_002877.2 1 836
GenBank mRNA AF493915.1 - 1 354
AL713766.1 - 1 459
BC012362.1 - 1 778 4650072
X52987.1 - 1 354

View Full Product Details