Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001185057.2 | NP_001171986.1 | 3 | 736 | |
NM_001276307.1 | NP_001263236.1 | 5 | 770 | ||
NM_001276309.1 | NP_001263238.1 | 3 | 630 | ||
NM_001276311.1 | NP_001263240.1 | 3 | 622 | ||
NM_001276312.1 | NP_001263241.1 | 5 | 1121 | ||
NM_001276319.1 | NP_001263248.1 | 5 | 1121 | ||
NM_003946.6 | NP_003937.1 | 3 | 746 | ||
XM_005256217.2 | XP_005256274.1 | 3 | 1373 | ||
XM_005256219.3 | XP_005256276.1 | 3 | 1259 | ||
XM_011523424.2 | XP_011521726.1 | 4 | 1216 | ||
XM_017023843.1 | XP_016879332.1 | 3 | 1383 | ||
GenBank mRNA | AF043244.1 | - | 3 | 663 | |
AF064599.1 | - | 3 | 587 | ||
AF064600.1 | - | 3 | 621 | ||
AK294145.1 | - | 5 | 1086 | ||
AK314899.1 | - | 3 | 651 | ||
BC012798.2 | - | 3 | 632 | 3506011 | |
BF568452.1 | - | 525 | |||
BQ685512.1 | - | 3 | 746 | ||
BT020158.1 | - | 2 | 544 |