Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001322321.1 | NP_001309250.1 | 13 | 1361 | |
NM_001322322.1 | NP_001309251.1 | 13 | 1361 | ||
NM_001322325.1 | NP_001309254.1 | 13 | 1361 | ||
NM_014636.2 | NP_055451.1 | 13 | 1324 | ||
XM_006717328.3 | XP_006717391.2 | 14 | 1335 | ||
XM_011519225.1 | XP_011517527.1 | 13 | 1284 | ||
XM_011519226.1 | XP_011517528.1 | 13 | 1284 | ||
XM_011519227.1 | XP_011517529.1 | 12 | 1233 | ||
XM_011519228.1 | XP_011517530.1 | 13 | 1284 | ||
XM_011519229.1 | XP_011517531.1 | 14 | 1341 | ||
XM_011519230.2 | XP_011517532.1 | 14 | 1335 | ||
XM_011519233.1 | XP_011517535.1 | 12 | 1122 | ||
XM_011519234.2 | XP_011517536.1 | 13 | 1418 | ||
XM_011519235.2 | XP_011517537.1 | 14 | 1509 | ||
XM_011519238.2 | XP_011517540.1 | 13 | 1284 | ||
XM_017015342.1 | XP_016870831.1 | 13 | 1284 | ||
XM_017015343.1 | XP_016870832.1 | 14 | 1328 | ||
XM_017015344.1 | XP_016870833.1 | 11 | 1101 | ||
XM_017015346.1 | XP_016870835.1 | 12 | 1122 | ||
XM_017015347.1 | XP_016870836.1 | 13 | 1181 | ||
XM_017015349.1 | XP_016870838.1 | 10 | 969 | ||
GenBank mRNA | AB002349.1 | - | 13 | 1324 |