Gene Transcripts

Gene Symbol
MOB4
Entrez Gene ID
25843
Gene Name
MOB family member 4, phocein
Gene Aliases
2C4D, CGI-95, MOB1, MOB3, MOBKL3, PHOCN, PREI3
Chromosome Location
Chr. 2: 197515571 - 197553699 on Build GRCh38
UniGene ID
Hs.633165
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100819.2 NP_001094289.1 7 558
NM_001204094.1 NP_001191023.1 8 575
NM_015387.4 NP_056202.2 8 621
NM_199482.3 NP_955776.1 8 631
GenBank mRNA AB015441.1 - 8 580
AF093825.1 - 8 596
AF151853.1 - 8 596
AF250319.1 - 8 606
AJ580638.1 - 8 577
AK027043.1 - 8 618
AK289377.1 - 8 611
AK292938.1 - 8 617
AK297294.1 - 492
AK297514.1 - 7 517
AK307497.1 - 7 489
AL080070.1 - 8 589
BC005237.1 - 8 621 3996585
CR457371.1 - 8 577
DC350915.1 - 7 554

Gene Symbol
HSPE1-MOB4
Entrez Gene ID
100529241
Gene Name
HSPE1-MOB4 readthrough
Gene Aliases
HSPE1-PHOCN
Chromosome Location
Chr. 2: 197499997 - 197553699 on Build GRCh38
UniGene ID
Hs.633165
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001202485.1 NP_001189414.1 9 1125

View Full Product Details