Gene Transcripts
- Gene Symbol
- ODF2L
- Entrez Gene ID
- 57489
- Gene Name
- outer dense fiber of sperm tails 2 like
- Gene Aliases
- dJ977L11.1
- Chromosome Location
- Chr. 1: 86346824 - 86396345 on Build GRCh38
- UniGene ID
- Hs.149360
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001007022.2 |
NP_001007023.2
|
6, 7 |
880 |
|
NM_001184765.1 |
NP_001171694.1
|
6, 7 |
887 |
|
NM_001184766.1 |
NP_001171695.1
|
6, 7 |
887 |
|
NM_020729.2 |
NP_065780.2
|
6, 7 |
887 |
|
XM_005271054.3 |
XP_005271111.1
|
6, 7 |
885 |
|
XM_011541815.2 |
XP_011540117.1
|
6, 7 |
885 |
|
XM_011541819.2 |
XP_011540121.1
|
6, 7 |
885 |
|
XM_011541821.2 |
XP_011540123.1
|
6, 7 |
885 |
|
XM_011541823.2 |
XP_011540125.1
|
6, 7 |
885 |
|
XM_011541824.2 |
XP_011540126.1
|
6, 7 |
885 |
|
XM_017001876.1 |
XP_016857365.1
|
6, 7 |
677 |
|
XM_017001877.1 |
XP_016857366.1
|
6, 7 |
677 |
|
XM_017001879.1 |
XP_016857368.1
|
6, 7 |
885 |
|
GenBank mRNA |
AB033055.1 |
- |
6, 7 |
800 |
|
AK302792.1 |
- |
6, 7 |
859 |
|
AK303209.1 |
- |
6, 7 |
884 |
|
AK308351.1 |
- |
3, 4 |
410 |
|
BC029420.1 |
- |
6, 7 |
605 |
|
BC047368.1 |
- |
6, 7 |
827 |
5276959 |
BC091490.1 |
- |
6, 7 |
786 |
30367572 |
BI768058.1 |
- |
5, 6 |
708 |
|
BX499763.1 |
- |
2, 3 |
101 |
|
View Full Product Details