Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032214.3 | NP_115590.1 | 8 | 1164 | |
NM_175077.2 | NP_778252.1 | 8 | 1114 | ||
GenBank mRNA | AF290985.1 | - | 7 | 741 | |
AF290986.1 | - | 7 | 691 | ||
AF326353.1 | - | 7 | 741 | ||
AK025645.1 | - | 8 | 801 | ||
AK291513.1 | - | 8 | 1144 | ||
BC042041.1 | - | 8 | 1103 | 4429896 | |
BQ052468.1 | - | 8 | 783 |