Gene Transcripts

Gene Symbol
PRIMA1
Entrez Gene ID
145270
Gene Name
proline rich membrane anchor 1
Gene Aliases
PRIMA
Chromosome Location
Chr. 14: 93718298 - 93789029 on Build GRCh38
UniGene ID
Hs.432401
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178013.3 NP_821092.1 4 601
XM_011536456.2 XP_011534758.1 5 1055
GenBank mRNA AY225516.1 - 4 601
AY225517.1 - 5 772

View Full Product Details