Gene Transcripts

Gene Symbol
PSORS1C2
Entrez Gene ID
170680
Gene Name
psoriasis susceptibility 1 candidate 2
Gene Aliases
C6orf17, SPR1
Chromosome Location
Chr. 6: 31137534 - 31139350 on Build GRCh38
UniGene ID
Hs.146824
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014069.2 NP_054788.2 2 545
GenBank mRNA AB031480.1 - 2 536
AF484420.1 - 2 545

View Full Product Details