Gene Transcripts

Gene Symbol
MTX3
Entrez Gene ID
345778
Gene Name
metaxin 3
Gene Aliases
Chromosome Location
Chr. 5: 79976716 - 79991267 on Build GRCh38
UniGene ID
Hs.531418
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010891.4 NP_001010891.4 1, 2 99
XM_005248495.3 XP_005248552.1 1, 2 101
GenBank mRNA AK296864.1 - 1, 2 94
BC150295.1 - 1, 2 89
BX647596.1 - 1, 2 96
DB456588.1 - 1, 2 99

View Full Product Details