Gene Transcripts

Gene Symbol
TSPAN33
Entrez Gene ID
340348
Gene Name
tetraspanin 33
Gene Aliases
PEN, PEN., TSPAN-33
Chromosome Location
Chr. 7: 129144716 - 129169694 on Build GRCh38
UniGene ID
Hs.27267
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178562.4 NP_848657.1 8 2882
GenBank mRNA AW089489.1 - 1 52

View Full Product Details