Gene Transcripts

Gene Symbol
OR56A4
Entrez Gene ID
120793
Gene Name
olfactory receptor family 56 subfamily A member 4
Gene Aliases
OR11-49
Chromosome Location
Chr. 11: 6002051 - 6003148 on Build GRCh38
UniGene ID
Hs.553571
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005179.2 NP_001005179.2 1 616
GenBank mRNA BC136986.1 - 1 662

View Full Product Details