Gene Transcripts

Gene Symbol
NLRP11
Entrez Gene ID
204801
Gene Name
NLR family pyrin domain containing 11
Gene Aliases
CLR19.6, NALP11, NOD17, PAN10, PYPAF6, PYPAF7
Chromosome Location
Chr. 19: 55785397 - 55836762 on Build GRCh38
UniGene ID
Hs.375039
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297743.1 NP_001284672.1 6 2249
NM_145007.3 NP_659444.2 9 3099
GenBank mRNA AK090621.1 - 4 1137
AK129784.1 - 4 1164
AK310394.1 - 11 3008
AY095145.1 - 7 2487
AY154466.1 - 6 2388
AY168967.1 - 7 2476
AY168968.1 - 6 2148
AY226382.1 - 6 2388
BC034730.1 - 7 2476 4341197

View Full Product Details