Gene Transcripts

Gene Symbol
OR10S1
Entrez Gene ID
219873
Gene Name
olfactory receptor family 10 subfamily S member 1
Gene Aliases
OR11-279
Chromosome Location
Chr. 11: 123976696 - 123977691 on Build GRCh38
UniGene ID
Hs.553637
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004474.1 NP_001004474.1 1 675
GenBank mRNA BC137037.1 - 1 765
BC137038.1 - 1 765 9021034

View Full Product Details