Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015140.3 | NP_055955.1 | 14 | 1982 | |
GenBank mRNA | AK123789.1 | - | 3 | 1051 | |
AK304464.1 | - | 4 | 468 | ||
BC001070.2 | - | 14 | 1946 | 3504490 | |
BE785973.1 | - | 1 | 81 | ||
CT841516.1 | - | 14 | 1923 | ||
D63487.1 | - | 14 | 1904 |