Gene Transcripts
- Gene Symbol
- SLC7A8
- Entrez Gene ID
- 23428
- Gene Name
- solute carrier family 7 member 8
- Gene Aliases
- LAT2, LPI-PC1
- Chromosome Location
- Chr. 14: 23125295 - 23183660 on Build GRCh38
- UniGene ID
- Hs.596643
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001267036.1 |
NP_001253965.1
|
2, 3 |
417 |
|
NM_012244.3 |
NP_036376.2
|
4, 5 |
1378 |
|
NM_182728.2 |
NP_877392.1
|
2, 3 |
316 |
|
NR_049767.1 |
|
2, 3 |
417 |
|
GenBank mRNA |
AB037669.1 |
- |
4, 5 |
1341 |
|
AF087908.1 |
- |
2, 3 |
271 |
|
AF087911.1 |
- |
3, 4 |
427 |
|
AF135828.1 |
- |
4, 5 |
853 |
|
AF171669.1 |
- |
4, 5 |
855 |
|
AK223610.1 |
- |
4, 5 |
1377 |
|
AK296702.1 |
- |
2, 3 |
370 |
|
AK307609.1 |
- |
2, 3 |
250 |
|
AK313465.1 |
- |
4, 5 |
632 |
|
AL365342.1 |
- |
4, 5 |
1363 |
|
AL365343.2 |
- |
4, 5 |
882 |
|
AL365414.1 |
- |
4, 5 |
882 |
|
BC036825.1 |
- |
2, 3 |
268 |
|
BC052250.1 |
- |
4, 5 |
661 |
6260264 |
BX248288.1 |
- |
2, 3 |
267 |
|
Y18483.1 |
- |
4, 5 |
1362 |
|
View Full Product Details