Gene Transcripts

Gene Symbol
OR5C1
Entrez Gene ID
392391
Gene Name
olfactory receptor family 5 subfamily C member 1
Gene Aliases
OR5C2P, OR9-31, OR9-F
Chromosome Location
Chr. 9: 122788933 - 122789895 on Build GRCh38
UniGene ID
Hs.534761
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001923.1 NP_001001923.1 1 420
GenBank mRNA BC136700.1 - 1 482
BC136701.1 - 1 482 9020691

View Full Product Details