Gene Transcripts

Gene Symbol
BORCS8-MEF2B
Entrez Gene ID
4207
Gene Name
BORCS8-MEF2B readthrough
Gene Aliases
LOC729991-MEF2B, MEF2B, MEF2BNB-MEF2B
Chromosome Location
Chr. 19: 19145567 - 19192591 on Build GRCh38
UniGene ID
Hs.153629
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005919.3 NP_005910.1 6 1056
NR_027307.1 8 1234
NR_027308.1 8 1234
GenBank mRNA AK301086.1 - 8 801
AK316328.1 - 8 801
BC126245.1 - 5 479 8991961
BC136457.1 - 4 403 9020446
BC171767.1 - 4 403 9054421
X63380.1 - 8 771
X68502.1 - 6 578

Gene Symbol
MEF2B
Entrez Gene ID
100271849
Gene Name
myocyte enhancer factor 2B
Gene Aliases
RSRFR2
Chromosome Location
Chr. 19: 19145567 - 19170289 on Build GRCh38
UniGene ID
Hs.153629
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145785.1 NP_001139257.1 4 443
GenBank mRNA AK057210.1 - 4 426
DA488022.1 - 5 561

View Full Product Details