Gene Transcripts

Gene Symbol
NKG7
Entrez Gene ID
4818
Gene Name
natural killer cell granule protein 7
Gene Aliases
GIG1, GMP-17, p15-TIA-1
Chromosome Location
Chr. 19: 51371616 - 51372706 on Build GRCh38
UniGene ID
Hs.10306
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005601.3 NP_005592.1 1, 2 326
GenBank mRNA BC015759.1 - 1, 2 255
S69115.1 - 1, 2 335

View Full Product Details