Gene Transcripts

Gene Symbol
SSUH2
Entrez Gene ID
51066
Gene Name
ssu-2 homolog (C. elegans)
Gene Aliases
C3orf32, SSU-2, fls485
Chromosome Location
Chr. 3: 8619400 - 8745035 on Build GRCh38
UniGene ID
Hs.736935
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256748.1 NP_001243677.1 5 399
NM_001256749.1 NP_001243678.1 5 420
NM_015931.2 NP_057015.1 5 566
NR_046358.1 6 647
XM_011533774.1 XP_011532076.1 13 2115
XM_017006510.1 XP_016861999.1 6 4880
XM_017006511.1 XP_016862000.1 12 1803
XM_017006512.1 XP_016862001.1 12 1972
XM_017006513.1 XP_016862002.1 11 1808
XM_017006514.1 XP_016862003.1 11 1714
XM_017006515.1 XP_016862004.1 13 2869
XM_017006516.1 XP_016862005.1 8 1230
XM_017006517.1 XP_016862006.1 8 1183
XM_017006518.1 XP_016862007.1 13 2094
XM_017006519.1 XP_016862008.1 6 4825
XM_017006520.1 XP_016862009.1 6 1176
XM_017006521.1 XP_016862010.1 9 1334
XM_017006522.1 XP_016862011.1 9 1225
XM_017006523.1 XP_016862012.1 7 928
XM_017006524.1 XP_016862013.1 8 1166
XM_017006525.1 XP_016862014.1 11 1735
XM_017006526.1 XP_016862015.1 6 814
XM_017006527.1 XP_016862016.1 12 1857
XM_017006528.1 XP_016862017.1 6 853
XM_017006529.1 XP_016862018.1 6 883
XM_017006530.1 XP_016862019.1 8 1316
XM_017006531.1 XP_016862020.1 7 1238
XM_017006532.1 XP_016862021.1 6 5738
XM_017006533.1 XP_016862022.1 12 1890
XM_017006534.1 XP_016862023.1 12 1836
XM_017006535.1 XP_016862024.1 6 6014
XM_017006536.1 XP_016862025.1 6 6017
XM_017006537.1 XP_016862026.1 6 6017
XM_017006538.1 XP_016862027.1 10 1433
XM_017006539.1 XP_016862028.1 5 545
XM_017006540.1 XP_016862029.1 6 724
XM_017006541.1 XP_016862030.1 3 366
XM_017006542.1 XP_016862031.1 3 344
GenBank mRNA AB024705.1 - 5 539
AK057972.1 - 6 643
AK093066.1 - 11 1596
AK122938.1 - 10 1381
AK225925.1 - 6 643
AK300175.1 - 5 399
BC052614.1 - 5 403 6300057
DB010779.1 - 5 420

View Full Product Details