Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164386.1 | NP_001157858.1 | 19 | 2986 | |
NM_001164387.1 | NP_001157859.1 | 20 | 3001 | ||
NM_001164388.1 | NP_001157860.1 | 19 | 2986 | ||
NM_001164389.1 | NP_001157861.1 | 19 | 2986 | ||
NM_016340.5 | NP_057424.3 | 19 | 2986 | ||
GenBank mRNA | AF394782.1 | - | 20 | 2977 | |
AF478468.1 | - | 19 | 2780 | ||
AF478469.1 | - | 19 | 2780 | ||
AF478567.1 | - | 19 | 2986 | ||
AK074318.1 | - | 11 | 1922 | ||
AK302640.1 | - | 19 | 2963 | ||
BC133703.1 | - | 19 | 2909 | 40124898 | |
BC140820.1 | - | 19 | 2861 | 9021688 | |
BC144627.1 | - | 19 | 2861 | 9053158 |