Gene Transcripts

Gene Symbol
MARCH5
Entrez Gene ID
54708
Gene Name
membrane associated ring-CH-type finger 5
Gene Aliases
MARCH-V, MITOL, RNF153
Chromosome Location
Chr. 10: 92291163 - 92353964 on Build GRCh38
UniGene ID
Hs.573490
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017824.4 NP_060294.1 5, 6 1056
XM_011539886.2 XP_011538188.1 6, 7 1204
GenBank mRNA AB191202.1 - 5, 6 812
AK000452.1 - 5, 6 1058
AK294799.1 - 5, 6 1130
BC015480.1 - 5, 6 1016 3905766

View Full Product Details