Gene Transcripts
- Gene Symbol
- LRTM2
- Entrez Gene ID
- 654429
- Gene Name
- leucine rich repeats and transmembrane domains 2
- Gene Aliases
- Chromosome Location
- Chr. 12: 1820267 - 1836753 on Build GRCh38
- UniGene ID
- Hs.585579
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039029.2 |
NP_001034118.1
|
3, 4 |
865 |
|
NM_001163925.1 |
NP_001157397.1
|
3, 4 |
901 |
|
NM_001163926.1 |
NP_001157398.1
|
3, 4 |
868 |
|
XM_011521015.1 |
XP_011519317.1
|
3, 4 |
1086 |
|
XM_017019848.1 |
XP_016875337.1
|
1, 2 |
273 |
|
GenBank mRNA |
AK095610.1 |
- |
3, 4 |
603 |
|
AK124687.1 |
- |
3, 4 |
472 |
|
BC129997.1 |
- |
2, 3 |
1090 |
|
BC146894.1 |
- |
3, 4 |
504 |
9056822 |
BC146910.1 |
- |
3, 4 |
504 |
9056838 |
BC150567.1 |
- |
2, 3 |
1090 |
|
BC150568.1 |
- |
1, 2 |
109 |
40127648 |
View Full Product Details