Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320458.1 | NP_001307387.1 | 2 | 314 | |
NM_003396.2 | NP_003387.1 | 2 | 314 | ||
XM_011525178.2 | XP_011523480.1 | 2 | 424 | ||
GenBank mRNA | AB063483.1 | - | 2 | 298 | |
AK127615.1 | - | 2 | 314 | ||
AY358217.1 | - | 2 | 334 | ||
BC064534.1 | - | 2 | 288 | 5588904 |