Gene Transcripts
- Gene Symbol
- NANOG
- Entrez Gene ID
- 79923
- Gene Name
- Nanog homeobox
- Gene Aliases
- Chromosome Location
- Chr. 12: 7789396 - 7796061 on Build GRCh38
- UniGene ID
- Hs.635882
- Species
- Homo sapiens
- Gene Symbol
- NANOGP1
- Entrez Gene ID
- 404635
- Gene Name
- Nanog homeobox pseudogene 1
- Gene Aliases
- NANOG2
- Chromosome Location
- Chr. 12: 7892356 - 7899246 on Build GRCh38
- UniGene ID
- Hs.635882
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK097770.1 |
- |
4 |
730 |
|
AY455283.1 |
- |
4 |
730 |
|
View Full Product Details