Gene Transcripts

Gene Symbol
TMEM88
Entrez Gene ID
92162
Gene Name
transmembrane protein 88
Gene Aliases
Chromosome Location
Chr. 17: 7854886 - 7856099 on Build GRCh38
UniGene ID
Hs.389669
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203411.1 NP_981956.1 1, 2 210
GenBank mRNA AK000032.1 - 1, 2 211
BC057812.1 - 1, 2 210 30332216

View Full Product Details