Gene Transcripts
- Gene Symbol
- MTRF1
- Entrez Gene ID
- 9617
- Gene Name
- mitochondrial translational release factor 1
- Gene Aliases
- MRF1, MTTRF1, RF1
- Chromosome Location
- Chr. 13: 41216369 - 41311794 on Build GRCh38
- UniGene ID
- Hs.382176
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004294.2 |
NP_004285.2
|
5, 6 |
797 |
|
XM_005266599.2 |
XP_005266656.1
|
7, 8 |
1046 |
|
XM_006719897.2 |
XP_006719960.1
|
7, 8 |
1154 |
|
XM_006719898.2 |
XP_006719961.1
|
7, 8 |
1138 |
|
XM_006719900.2 |
XP_006719963.1
|
6, 7 |
867 |
|
XM_011535315.2 |
XP_011533617.1
|
7, 8 |
1263 |
|
XM_011535317.2 |
XP_011533619.1
|
6, 7 |
1146 |
|
XM_011535318.2 |
XP_011533620.1
|
7, 8 |
1306 |
|
XM_011535320.2 |
XP_011533622.1
|
7, 8 |
1242 |
|
XM_011535321.2 |
XP_011533623.1
|
10, 11 |
1378 |
|
XM_011535323.2 |
XP_011533625.1
|
7, 8 |
1108 |
|
XM_011535325.2 |
XP_011533627.1
|
6, 7 |
1061 |
|
XM_011535326.2 |
XP_011533628.1
|
6, 7 |
1107 |
|
XM_017020856.1 |
XP_016876345.1
|
6, 7 |
1107 |
|
XM_017020857.1 |
XP_016876346.1
|
9, 10 |
1241 |
|
XM_017020858.1 |
XP_016876347.1
|
8, 9 |
1236 |
|
XM_017020859.1 |
XP_016876348.1
|
6, 7 |
1107 |
|
XM_017020860.1 |
XP_016876349.1
|
8, 9 |
1124 |
|
XM_017020861.1 |
XP_016876350.1
|
8, 9 |
1161 |
|
XM_017020862.1 |
XP_016876351.1
|
7, 8 |
2319 |
|
XM_017020863.1 |
XP_016876352.1
|
7, 8 |
2167 |
|
XM_017020864.1 |
XP_016876353.1
|
7, 8 |
2059 |
|
XM_017020865.1 |
XP_016876354.1
|
6, 7 |
2011 |
|
XM_017020866.1 |
XP_016876355.1
|
5, 6 |
1876 |
|
XM_017020867.1 |
XP_016876356.1
|
7, 8 |
1024 |
|
XM_017020868.1 |
XP_016876357.1
|
5, 6 |
992 |
|
XM_017020869.1 |
XP_016876358.1
|
6, 7 |
890 |
|
XM_017020870.1 |
XP_016876359.1
|
7, 8 |
1154 |
|
XM_017020871.1 |
XP_016876360.1
|
6, 7 |
867 |
|
XM_017020872.1 |
XP_016876361.1
|
6, 7 |
1107 |
|
XM_017020873.1 |
XP_016876362.1
|
10, 11 |
1448 |
|
XM_017020874.1 |
XP_016876363.1
|
7, 8 |
1000 |
|
XM_017020875.1 |
XP_016876364.1
|
7, 8 |
994 |
|
XM_017020876.1 |
XP_016876365.1
|
6, 7 |
2334 |
|
XM_017020877.1 |
XP_016876366.1
|
8, 9 |
1169 |
|
XM_017020878.1 |
XP_016876367.1
|
7, 8 |
1046 |
|
GenBank mRNA |
AF072934.1 |
- |
5, 6 |
768 |
|
AK294343.1 |
- |
6, 7 |
954 |
|
BC042196.1 |
- |
5, 6 |
797 |
5198308 |
View Full Product Details