Gene Transcripts

Gene Symbol
SNX17
Entrez Gene ID
9784
Gene Name
sorting nexin 17
Gene Aliases
Chromosome Location
Chr. 2: 27370496 - 27377533 on Build GRCh38
UniGene ID
Hs.278569
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267059.1 NP_001253988.1 5 601
NM_001267060.1 NP_001253989.1 4 562
NM_001267061.1 NP_001253990.1 5 472
NM_014748.3 NP_055563.1 5 637
NR_049782.1 5 762
NR_049783.1 6 735
NR_049784.1 6 711
NR_049785.1 4 644
NR_049786.1 5 593
NR_049787.1 3 444
NR_049788.1 3 444
XM_011533203.1 XP_011531505.1 4 411
XM_017005405.1 XP_016860894.1 4 411
GenBank mRNA AK129689.1 - 5 521
AK222543.1 - 5 520
AK293252.1 - 6 611
AK293464.1 - 5 453
AK295278.1 - 3 323
AK297054.1 - 5 646
AK298620.1 - 5 472
AK298856.1 - 5 474
AK298869.1 - 4 423
AK300144.1 - 5 484
AK300641.1 - 3 317
AK307122.1 - 4 520
AK308487.1 - 4 523
AK309640.1 - 4 395
AK316373.1 - 5 416
BC002524.1 - 5 498 3140610
BC002610.1 - 5 487 3143104
BC014620.2 - 5 498 3010648
BC050590.1 - 5 492 6044270
BT007167.1 - 5 389
CR457081.1 - 5 389
D31764.1 - 5 611

View Full Product Details