Gene Transcripts

Gene Symbol
Bend7
Entrez Gene ID
209645
Gene Name
BEN domain containing 7
Gene Aliases
1110017O21Rik, E130319B15Rik
Chromosome Location
Chr. 2: 4717831 - 4802146 on Build GRCm38
UniGene ID
Mm.149539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178663.4 NP_848778.1 7, 8 1600
GenBank mRNA AK030428.1 - 7, 8 1287
BC056476.1 - 4, 5 644
BC119370.1 - 7, 8 1239 8734119

View Full Product Details