Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145955.3 | NP_666067.1 | 10 | 1329 | |
GenBank mRNA | AK083385.1 | - | 10 | 1390 | |
AK189603.1 | - | 2 | 191 | ||
BC025641.1 | - | 10 | 1359 | 5322565 | |
BC038342.1 | - | 10 | 1329 | 4191750 |