Gene Transcripts

Gene Symbol
Vipr1
Entrez Gene ID
22354
Gene Name
vasoactive intestinal peptide receptor 1
Gene Aliases
AV071699, VIP-R1, VPAC1
Chromosome Location
Chr. 9: 121642716 - 121672954 on Build GRCm38
UniGene ID
Mm.282007
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011703.4 NP_035833.2 10 1122
GenBank mRNA AF266282.1 - 10 1004
AK052465.1 - 10 1123

View Full Product Details