Gene Transcripts

Gene Symbol
Nyx
Entrez Gene ID
236690
Gene Name
nyctalopin
Gene Aliases
CLNP, CSNB1, CSNB4, nob
Chromosome Location
Chr. X: 13461095 - 13489314 on Build GRCm38
UniGene ID
Mm.90233
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173415.4 NP_775591.1 3 489
GenBank mRNA AY114303.1 - 2189
BC035244.1 - 3 489 5362608
BC048381.1 - 3 488 5362608

View Full Product Details