Gene Transcripts

Gene Symbol
Med19
Entrez Gene ID
381379
Gene Name
mediator complex subunit 19
Gene Aliases
2410018M14Rik, 3110040A13Rik, LCMR1
Chromosome Location
Chr. 2: 84678402 - 84688215 on Build GRCm38
UniGene ID
Mm.219643
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025885.3 NP_080161.1 2 484
GenBank mRNA AK005399.1 - 2 452
AK010552.1 - 2 462
AK160579.1 - 2 450
AK168787.1 - 2 452
BB611514.1 - 2 484
BC025475.1 - 1 58
BC044207.1 - 2 346
BC044302.1 - 2 346
BC055404.1 - 2 439
BC082559.1 - 2 449 30606373

View Full Product Details