Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172253.2 | NP_758457.1 | 2 | 338 | |
GenBank mRNA | AK008677.1 | - | 2 | 320 | |
AK010829.1 | - | 2 | 339 | ||
AK016638.1 | - | 2 | 335 | ||
AK017016.1 | - | 2 | 328 | ||
AK017810.1 | - | 2 | 323 | ||
AK153090.1 | - | 328 | |||
AK160682.1 | - | 2 | 326 | ||
AK179512.1 | - | 2 | 94 | ||
BC082580.1 | - | 310 | |||
BC139162.1 | - | 2 | 326 | 8862184 | |
BC139163.1 | - | 2 | 326 | 8862185 |