Gene Transcripts
- Gene Symbol
- Pmm2
- Entrez Gene ID
- 54128
- Gene Name
- phosphomannomutase 2
- Gene Aliases
- AI585868, C86848
- Chromosome Location
- Chr. 16: 8637707 - 8657552 on Build GRCm38
- UniGene ID
- Mm.9699
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016881.2 |
NP_058577.1
|
7, 8 |
704 |
|
GenBank mRNA |
AF043514.1 |
- |
7, 8 |
704 |
|
AK002858.1 |
- |
7, 8 |
646 |
|
AK003334.1 |
- |
7, 8 |
662 |
|
AK005074.1 |
- |
7, 8 |
686 |
|
AK134850.1 |
- |
|
656 |
|
AK145402.1 |
- |
7, 8 |
656 |
|
AK149116.1 |
- |
7, 8 |
666 |
|
BC037101.1 |
- |
7, 8 |
681 |
|
BC046325.1 |
- |
7, 8 |
652 |
6395635 |
BC094448.1 |
- |
7, 8 |
670 |
4238664 |
View Full Product Details