Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026742.4 | NP_081018.1 | 1 | 108 | |
GenBank mRNA | AK003553.2 | - | 1 | 63 | |
AK019042.1 | - | 1 | 85 | ||
AK034728.1 | - | 1 | 91 | ||
AK160341.1 | - | 1 | 104 | ||
BC028764.1 | - | 1 | 77 | 1195846 | |
BY163651.1 | - | 1 | 113 |