Gene Transcripts

Gene Symbol
Mdk
Entrez Gene ID
17242
Gene Name
midkine
Gene Aliases
MK, Mek
Chromosome Location
Chr. 2: 91929805 - 91932297 on Build GRCm38
UniGene ID
Mm.906
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012335.2 NP_001012335.1 3, 4 402
NM_001012336.2 NP_001012336.1 3, 4 399
NM_001291481.1 NP_001278410.1 3, 4 309
NM_001291482.1 NP_001278411.1 2, 3 322
NM_010784.5 NP_034914.1 3, 4 601
GenBank mRNA AK032742.1 - 3, 4 305
AK084457.1 - 3, 4 322
AL364704.1 - 3, 4 282
BC012244.1 - 3, 4 270 4167496
CF728526.1 - 3, 4 295
CF731708.1 - 3, 4 398
CF738400.1 - 3, 4 407
CF740555.1 - 1, 2 119
CJ174056.1 - 3, 4 410
CJ174578.1 - 2, 3 197
M19662.1 - 2, 3 480
M34327.1 - 3, 4 275
M34328.1 - 3, 4 586
M35833.1 - 3, 4 275

View Full Product Details