Gene Transcripts
- Gene Symbol
- Prkacb
- Entrez Gene ID
- 18749
- Gene Name
- protein kinase, cAMP dependent, catalytic, beta
- Gene Aliases
- Pkacb
- Chromosome Location
- Chr. 3: 146729579 - 146812946 on Build GRCm38
- UniGene ID
- Mm.16766
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164198.1 |
NP_001157670.1
|
9, 10 |
1193 |
|
NM_001164199.1 |
NP_001157671.1
|
9, 10 |
1066 |
|
NM_001164200.1 |
NP_001157672.1
|
9, 10 |
1158 |
|
NM_011100.4 |
NP_035230.1
|
9, 10 |
1197 |
|
GenBank mRNA |
AK035560.1 |
- |
2, 3 |
281 |
|
AK048319.1 |
- |
9, 10 |
1189 |
|
AK081984.1 |
- |
2, 3 |
207 |
|
AK139419.1 |
- |
9, 10 |
938 |
|
AK148728.1 |
- |
9, 10 |
1007 |
|
AK150122.1 |
- |
9, 10 |
1159 |
|
AK150134.1 |
- |
9, 10 |
1159 |
|
AK163585.1 |
- |
9, 10 |
971 |
|
BC054533.1 |
- |
9, 10 |
1181 |
5704796 |
J02626.1 |
- |
9, 10 |
1126 |
|
X61434.1 |
- |
1, 2 |
96 |
|
View Full Product Details