Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153388.4 | NP_700437.2 | 2 | 1886 | |
GenBank mRNA | AK081560.1 | - | 1 | 2169 | |
AK163603.1 | - | 2 | 1744 | ||
AK214339.1 | - | 1 | 290 | ||
BC023036.1 | - | 1 | 1499 | 5358628 | |
BC023156.1 | - | 2 | 1542 | 4951386 | |
DQ078786.1 | - | 1 | 1171 |