Gene Transcripts

Gene Symbol
Phldb3
Entrez Gene ID
232970
Gene Name
pleckstrin homology-like domain, family B, member 3
Gene Aliases
EG232970, ENSMUSG00000061511, Gm10102
Chromosome Location
Chr. 7: 24610272 - 24629297 on Build GRCm38
UniGene ID
Mm.333663
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102613.1 NP_001096083.1 13, 14 1750

View Full Product Details