Gene Transcripts

Gene Symbol
Ppp2r2c
Entrez Gene ID
269643
Gene Name
protein phosphatase 2, regulatory subunit B, gamma
Gene Aliases
6330548O06Rik, IMYPNO, IMYPNO1, PR52
Chromosome Location
Chr. 5: 36866843 - 36955078 on Build GRCm38
UniGene ID
Mm.41694
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172994.2 NP_766582.1 3 490
GenBank mRNA AK032023.1 - 3 393
AK049830.1 - 3 376
AY348866.1 - 3 492
BC059811.1 - 3 490 6849535

View Full Product Details