Gene Transcripts
- Gene Symbol
- Mug2
- Entrez Gene ID
- 17837
- Gene Name
- murinoglobulin 2
- Gene Aliases
- Cpamd8
- Chromosome Location
- Chr. 6: 122006719 - 122085968 on Build GRCm38
- UniGene ID
- Mm.479223
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008646.3 |
NP_032672.2
|
12 |
1409 |
|
GenBank mRNA |
M65238.1 |
- |
12 |
1412 |
|
View Full Product Details