Gene Transcripts

Gene Symbol
Myl2
Entrez Gene ID
17906
Gene Name
myosin, light polypeptide 2, regulatory, cardiac, slow
Gene Aliases
MLC-2, MLC-2v, Mlc2v, Mylpc
Chromosome Location
Chr. 5: 122100904 - 122113396 on Build GRCm38
UniGene ID
Mm.1529
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010861.3 NP_034991.3 3, 4 212
GenBank mRNA AK002367.1 - 3, 4 213
AK146674.1 - 3, 4 216
BC061144.1 - 213 30301284
M91602.1 - 3, 4 204

View Full Product Details