Gene Transcripts

Gene Symbol
Olfr12
Entrez Gene ID
257890
Gene Name
olfactory receptor 12
Gene Aliases
MOR208-5
Chromosome Location
Chr. 1: 92619881 - 92621001 on Build GRCm38
UniGene ID
Mm.337259
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206896.2 NP_996779.1 1 628
GenBank mRNA BC120559.1 - 1 628
BC120561.1 - 1 628 40129484

View Full Product Details