Gene Transcripts

Gene Symbol
Trip13
Entrez Gene ID
69716
Gene Name
thyroid hormone receptor interactor 13
Gene Aliases
2410002G23Rik, D13Ertd328e
Chromosome Location
Chr. 13: 73912462 - 73937767 on Build GRCm38
UniGene ID
Mm.275095
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027182.2 NP_081458.1 12 1370
GenBank mRNA AK010336.1 - 12 1332
AK142463.1 - 1343
AK146877.1 - 12 1371
AK151568.1 - 12 1306
BC023834.1 - 6 578
BC064315.1 - 12 1293
BC126946.1 - 12 1269 40090292

View Full Product Details