Gene Transcripts

Gene Symbol
Prr5
Entrez Gene ID
109270
Gene Name
proline rich 5 (renal)
Gene Aliases
AU043908, Arhgap8, C030017C09Rik, C78947, Protor-1, Protor1
Chromosome Location
Chr. 15: 84663171 - 84706409 on Build GRCm38
UniGene ID
Mm.291372
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146061.4 NP_666173.4 5 544
GenBank mRNA AF482997.1 - 5 539
AK046057.1 - 6 361
AK081231.1 - 5 540
BC024991.1 - 6 428 5065129

View Full Product Details