Gene Transcripts

Gene Symbol
Mcpt4
Entrez Gene ID
17227
Gene Name
mast cell protease 4
Gene Aliases
MMCP-4, MMCP-4A, MMCP-4B, Mcp-4, Mcp4
Chromosome Location
Chr. 14: 56059744 - 56062310 on Build GRCm38
UniGene ID
Mm.439684
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010779.2 NP_034909.2 4 431
GenBank mRNA AB051900.1 - 4 379
AK144385.1 - 4 432
AY007569.1 - 4 431
BC026198.1 - 4 407 1314753
CJ046459.1 - 4 434
M55617.1 - 431
X68804.1 - 4 378

View Full Product Details