Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008675.2 | NP_032701.2 | 3 | 399 | |
GenBank mRNA | AK133787.1 | - | 3 | 398 | |
AK146535.1 | - | 3 | 413 | ||
AK158008.1 | - | 3 | 406 | ||
AK167311.1 | - | 3 | 397 | ||
BC099856.1 | - | 3 | 277 | 40039759 | |
BC099857.1 | - | 3 | 277 | 40039762 | |
BC099858.1 | - | 3 | 277 | 40039757 | |
BC107810.1 | - | 2 | 263 | 40039758 | |
D50263.1 | - | 3 | 294 |