Gene Transcripts

Gene Symbol
Nsg2
Entrez Gene ID
18197
Gene Name
neuron specific gene family member 2
Gene Aliases
Chromosome Location
Chr. 11: 32000386 - 32059211 on Build GRCm38
UniGene ID
Mm.3304
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290680.1 NP_001277609.1 4, 5 377
NM_001290681.1 NP_001277610.1 4, 5 373
NM_008741.4 NP_032767.1 4, 5 513
GenBank mRNA AK011563.1 - 3, 4 329
AK075612.1 - 4, 5 470
AK088499.1 - 4, 5 505
BC018224.1 - 4, 5 445 4527900
BU058771.1 - 4, 5 376
DN176718.1 - 4, 5 513
U17259.1 - 4, 5 473

View Full Product Details