Gene Transcripts

Gene Symbol
St8sia4
Entrez Gene ID
20452
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4
Gene Aliases
PST, PST-1, SIAT8-D, ST8SiaIV, Siat8d
Chromosome Location
Chr. 1: 95587682 - 95667649 on Build GRCm38
UniGene ID
Mm.306228
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159745.1 NP_001153217.1 3 620
NM_009183.2 NP_033209.1 3 620
GenBank mRNA AJ223956.1 - 3 611
AK003690.1 - 3 562
AK041723.1 - 3 553
BC060112.1 - 3 597 6405078
X86000.1 - 620

View Full Product Details