Gene Transcripts

Gene Symbol
Alyref
Entrez Gene ID
21681
Gene Name
Aly/REF export factor
Gene Aliases
ALY, REF1, Refbp1, Thoc4
Chromosome Location
Chr. 11: 120594516 - 120598436 on Build GRCm38
UniGene ID
Mm.1886
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011568.1 NP_035698.1 1, 2 258
GenBank mRNA BC120588.1 - 1, 2 350
BC137658.1 - 1, 2 350 8860677
U89876.1 - 1, 2 258

View Full Product Details